Anti-TYMP Mouse Monoclonal Antibody [clone: 1E7]

Toimittaja: OriGene
ORIGCF800959EA 749 EUR
ORIGCF800959
Anti-TYMP Mouse Monoclonal Antibody [clone: 1E7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: TYMP
Clonality: Monoclonal
Clone: 1E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR