Anti-TYMS Mouse Monoclonal Antibody [clone: 1E12]

Toimittaja: OriGene
ORIGCF801764EA 749 EUR
ORIGCF801764
Anti-TYMS Mouse Monoclonal Antibody [clone: 1E12]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: TYMS
Clonality: Monoclonal
Clone: 1E12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR