Anti-DOCK8 Mouse Monoclonal Antibody [clone: 7H2]

Toimittaja: OriGene
ORIGCF801822EA 749 EUR
ORIGCF801822
Anti-DOCK8 Mouse Monoclonal Antibody [clone: 7H2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: DOCK8
Clonality: Monoclonal
Clone: 7H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR