Anti-SAP30BP Mouse Monoclonal Antibody [clone: 6E10]

Toimittaja: OriGene
ORIGCF803106EA 749 EUR
ORIGCF803106
Anti-SAP30BP Mouse Monoclonal Antibody [clone: 6E10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SAP30BP
Clonality: Monoclonal
Clone: 6E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR