Anti-PRRX1 Mouse Monoclonal Antibody [clone: 2E8]

Toimittaja: OriGene
ORIGCF803117EA 749 EUR
ORIGCF803117
Anti-PRRX1 Mouse Monoclonal Antibody [clone: 2E8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PRRX1
Clonality: Monoclonal
Clone: 2E8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR