Anti-ABCB1 Mouse Monoclonal Antibody [clone: 5B3]

Toimittaja: OriGene
ORIGCF801055EA 749 EUR
ORIGCF801055
Anti-ABCB1 Mouse Monoclonal Antibody [clone: 5B3]
Vasta-aineet
Recommended Dilutions: Immunochemistry: 1:150

Type: Primary
Antigen: ABCB1
Clonality: Monoclonal
Clone: 5B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR