Anti-PIK3CD Mouse Monoclonal Antibody [clone: 2H3]

Toimittaja: OriGene
ORIGCF801891EA 749 EUR
ORIGCF801891
Anti-PIK3CD Mouse Monoclonal Antibody [clone: 2H3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150

Type: Primary
Antigen: PIK3CD
Clonality: Monoclonal
Clone: 2H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR