Anti-VSNL1 Mouse Monoclonal Antibody [clone: 1F7]

Toimittaja: OriGene
ORIGCF801896EA 749 EUR
ORIGCF801896
Anti-VSNL1 Mouse Monoclonal Antibody [clone: 1F7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: VSNL1
Clonality: Monoclonal
Clone: 1F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR