Anti-CEACAM5 Mouse Monoclonal Antibody [clone: 2B12]

Toimittaja: OriGene
ORIGCF803181EA 749 EUR
ORIGCF803181
Anti-CEACAM5 Mouse Monoclonal Antibody [clone: 2B12]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: CEACAM5
Clonality: Monoclonal
Clone: 2B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR