Anti-ZNF365 Mouse Monoclonal Antibody [clone: 3B1]

Toimittaja: OriGene
ORIGCF800111EA 749 EUR
ORIGCF800111
Anti-ZNF365 Mouse Monoclonal Antibody [clone: 3B1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: ZNF365
Clonality: Monoclonal
Clone: 3B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR