Anti-MUC16 Mouse Monoclonal Antibody [clone: 2F1]

Toimittaja: OriGene
ORIGCF801212EA 770 EUR
ORIGCF801212
Anti-MUC16 Mouse Monoclonal Antibody [clone: 2F1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: MUC16
Clonality: Monoclonal
Clone: 2F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR