Anti-PAX5 Mouse Monoclonal Antibody [clone: 4F9]

Toimittaja: OriGene
ORIGCF801969EA 749 EUR
ORIGCF801969
Anti-PAX5 Mouse Monoclonal Antibody [clone: 4F9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: PAX5
Clonality: Monoclonal
Clone: 4F9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR