Anti-EGFR Mouse Monoclonal Antibody [clone: 3F2]

Toimittaja: OriGene
ORIGCF802016EA 749 EUR
ORIGCF802016
Anti-EGFR Mouse Monoclonal Antibody [clone: 3F2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: 3F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR