Anti-FOXI1 Mouse Monoclonal Antibody [clone: 1H9]

Toimittaja: OriGene
ORIGCF800147EA 749 EUR
ORIGCF800147
Anti-FOXI1 Mouse Monoclonal Antibody [clone: 1H9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: FOXI1
Clonality: Monoclonal
Clone: 1H9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR