Anti-VSNL1 Mouse Monoclonal Antibody [clone: 6A5]

Toimittaja: OriGene
ORIGCF802024EA 749 EUR
ORIGCF802024
Anti-VSNL1 Mouse Monoclonal Antibody [clone: 6A5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: VSNL1
Clonality: Monoclonal
Clone: 6A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR