Anti-ETV7 Mouse Monoclonal Antibody [clone: 2A10]

Toimittaja: OriGene
ORIGCF803463EA 749 EUR
ORIGCF803463
Anti-ETV7 Mouse Monoclonal Antibody [clone: 2A10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ETV7
Clonality: Monoclonal
Clone: 2A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR