Anti-ZNF286A Mouse Monoclonal Antibody [clone: 5E11]

Toimittaja: OriGene
ORIGCF803481EA 770 EUR
ORIGCF803481
Anti-ZNF286A Mouse Monoclonal Antibody [clone: 5E11]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ZNF286A
Clonality: Monoclonal
Clone: 5E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR