Anti-ALK Mouse Monoclonal Antibody [clone: 11H4]

Toimittaja: OriGene
ORIGCF801324EA 749 EUR
ORIGCF801324
Anti-ALK Mouse Monoclonal Antibody [clone: 11H4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ALK
Clonality: Monoclonal
Clone: 11H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR