Anti-VIM Mouse Monoclonal Antibody [clone: 6B12]

Toimittaja: OriGene
ORIGCF801326EA 749 EUR
ORIGCF801326
Anti-VIM Mouse Monoclonal Antibody [clone: 6B12]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: 6B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR