Anti-KI67 Rat Monoclonal Antibody [clone: 6C12]

Toimittaja: OriGene
ORIGCF802095EA 749 EUR
ORIGCF802095
Anti-KI67 Rat Monoclonal Antibody [clone: 6C12]
Vasta-aineet
Recommended Dilutions: Immunochemistry: 1:150

Type: Primary
Antigen: KI67
Clonality: Monoclonal
Clone: 6C12
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR