Anti-NBN Mouse Monoclonal Antibody [clone: 2E10]

Toimittaja: OriGene
ORIGCF801427EA 749 EUR
ORIGCF801427
Anti-NBN Mouse Monoclonal Antibody [clone: 2E10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: NBN
Clonality: Monoclonal
Clone: 2E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR