Anti-PIK3CA Mouse Monoclonal Antibody [clone: 6D1]

Toimittaja: OriGene
ORIGCF801483EA 749 EUR
ORIGCF801483
Anti-PIK3CA Mouse Monoclonal Antibody [clone: 6D1]
Vasta-aineet
Recommended Dilutions: Immunochemistry: 1:150

Type: Primary
Antigen: PIK3CA
Clonality: Monoclonal
Clone: 6D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR