Anti-IL1RN Mouse Monoclonal Antibody [clone: 2B1]

Toimittaja: OriGene
ORIGCF803422EA 749 EUR
ORIGCF803422
Anti-IL1RN Mouse Monoclonal Antibody [clone: 2B1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: IL1RN
Clonality: Monoclonal
Clone: 2B1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR