Anti-AGTPBP1 Mouse Monoclonal Antibody [clone: 9C8]

Toimittaja: OriGene
ORIGCF800287EA 749 EUR
ORIGCF800287
Anti-AGTPBP1 Mouse Monoclonal Antibody [clone: 9C8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: AGTPBP1
Clonality: Monoclonal
Clone: 9C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR