Anti-TLE1 Mouse Monoclonal Antibody [clone: 1D6]

Toimittaja: OriGene
ORIGCF800301EA 749 EUR
ORIGCF800301
Anti-TLE1 Mouse Monoclonal Antibody [clone: 1D6]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TLE1
Clonality: Monoclonal
Clone: 1D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR