Anti-F13A1 Mouse Monoclonal Antibody [clone: 6F5]

Toimittaja: OriGene
ORIGCF800352EA 770 EUR
ORIGCF800352
Anti-F13A1 Mouse Monoclonal Antibody [clone: 6F5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: F13A1
Clonality: Monoclonal
Clone: 6F5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR