Anti-WWC1 Mouse Monoclonal Antibody [clone: 1B1]

Toimittaja: OriGene
ORIGCF801574EA 749 EUR
ORIGCF801574
Anti-WWC1 Mouse Monoclonal Antibody [clone: 1B1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: WWC1
Clonality: Monoclonal
Clone: 1B1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR