Anti-KLK8 Mouse Monoclonal Antibody [clone: 3H1]

Toimittaja: OriGene
ORIGCF801586EA 749 EUR
ORIGCF801586
Anti-KLK8 Mouse Monoclonal Antibody [clone: 3H1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: KLK8
Clonality: Monoclonal
Clone: 3H1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR