Anti-NDUFS2 Mouse Monoclonal Antibody [clone: 2G2]

Toimittaja: OriGene
ORIGCF802325EA 770 EUR
ORIGCF802325
Anti-NDUFS2 Mouse Monoclonal Antibody [clone: 2G2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: NDUFS2
Clonality: Monoclonal
Clone: 2G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR