Anti-MSMB Mouse Monoclonal Antibody [clone: 2C11]

Toimittaja: OriGene
ORIGCF803521EA 749 EUR
ORIGCF803521
Anti-MSMB Mouse Monoclonal Antibody [clone: 2C11]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-1:1000

Type: Primary
Antigen: MSMB
Clonality: Monoclonal
Clone: 2C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR