Anti-KLHL12 Mouse Monoclonal Antibody [clone: 1D2]

Toimittaja: OriGene
ORIGCF803564EA 749 EUR
ORIGCF803564
Anti-KLHL12 Mouse Monoclonal Antibody [clone: 1D2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: KLHL12
Clonality: Monoclonal
Clone: 1D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR