Anti-EPHX1 Mouse Monoclonal Antibody [clone: 4G4]

Toimittaja: OriGene
ORIGCF800409EA 749 EUR
ORIGCF800409
Anti-EPHX1 Mouse Monoclonal Antibody [clone: 4G4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: EPHX1
Clonality: Monoclonal
Clone: 4G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR