Anti-PTPRC Mouse Monoclonal Antibody [clone: 2E7]

Toimittaja: OriGene
ORIGCF506046EA 749 EUR
ORIGCF506046
Anti-PTPRC Mouse Monoclonal Antibody [clone: 2E7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:400-4000; Immunochemistry: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:50

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: 2E7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR