Anti-PIK3C2B Mouse Monoclonal Antibody [clone: 3B1]

Toimittaja: OriGene
ORIGCF506052EA 770 EUR
ORIGCF506052
Anti-PIK3C2B Mouse Monoclonal Antibody [clone: 3B1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150

Type: Primary
Antigen: PIK3C2B
Clonality: Monoclonal
Clone: 3B1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR