Anti-ERG Mouse Monoclonal Antibody [clone: 6A4]

Toimittaja: OriGene
ORIGCF506078EA 749 EUR
ORIGCF506078
Anti-ERG Mouse Monoclonal Antibody [clone: 6A4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: ERG
Clonality: Monoclonal
Clone: 6A4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR