Anti-ERG Mouse Monoclonal Antibody [clone: 5D10]

Toimittaja: OriGene
ORIGCF506079EA 749 EUR
ORIGCF506079
Anti-ERG Mouse Monoclonal Antibody [clone: 5D10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-4000; Immunofluorescence: 1:100

Type: Primary
Antigen: ERG
Clonality: Monoclonal
Clone: 5D10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR