Anti-CD5 Mouse Monoclonal Antibody [clone: 1A8]

Toimittaja: OriGene
ORIGCF802403EA 749 EUR
ORIGCF802403
Anti-CD5 Mouse Monoclonal Antibody [clone: 1A8]
Vasta-aineet
Recommended Dilutions: Flow Cytometry: 1:50

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 1A8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR