Anti-CHAF1B Mouse Monoclonal Antibody [clone: 4F7]

Toimittaja: OriGene
ORIGCF506840EA 749 EUR
ORIGCF506840
Anti-CHAF1B Mouse Monoclonal Antibody [clone: 4F7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-1000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: CHAF1B
Clonality: Monoclonal
Clone: 4F7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR