Anti-A4GNT Mouse Monoclonal Antibody [clone: 1B10]

Toimittaja: OriGene
ORIGCF800432EA 770 EUR
ORIGCF800432
Anti-A4GNT Mouse Monoclonal Antibody [clone: 1B10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: A4GNT
Clonality: Monoclonal
Clone: 1B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR