Anti-DCX Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA323627EA 564 EUR
ORIGTA323627
Anti-DCX Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1000

Type: Primary
Antigen: DCX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR