Anti-MAP7 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA590643EA 684 EUR
ORIGTA590643
Anti-MAP7 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot 1:5000-20000, ELISA 1:100-1:2000

Type: Primary
Antigen: MAP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR