Anti-TNFRSF8 Mouse Monoclonal Antibody [clone: 2G7]

Toimittaja: OriGene
ORIGTA804021EA 616 EUR
ORIGTA804021
Anti-TNFRSF8 Mouse Monoclonal Antibody [clone: 2G7]
Vasta-aineet
Recommended Dilutions: Immunochemistry: 1:150; Flow Cytometry: 1:50

Type: Primary
Antigen: TNFRSF8
Clonality: Monoclonal
Clone: 2G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR