Anti-6xHis Mouse Monoclonal Antibody (Biotin) [clone: 14A231]

Toimittaja: Rockland Immunochemicals
ROCK200-306-382EA 752 EUR
ROCK200-306-382
Anti-6xHis Mouse Monoclonal Antibody (Biotin) [clone: 14A231]
Vasta-aineet
Type: Primary
Antigen: 6xHis
Clonality: Monoclonal
Clone: 14A231
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR