Anti-OPTN Rabbit Polyclonal Antibody

Toimittaja: Cayman Chemical
CAYM100000-1EA 544 EUR
CAYM100000-1
Anti-OPTN Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: OPTN
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR