Anti-TPH Rabbit Polyclonal Antibody

Toimittaja: Cayman Chemical
CAYM10009398-1EA 591 EUR
CAYM10009398-1
Anti-TPH Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: TPH
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR