Anti-MAPKK1 Rabbit Polyclonal Antibody

Toimittaja: Cayman Chemical
CAYM10009518-1EA 726 EUR
CAYM10009518-1
Anti-MAPKK1 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: MAPKK1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR