Anti-FABP3 Mouse Monoclonal Antibody [clone: CC68]

Toimittaja: Cayman Chemical
CAYM10233-1EA 532 EUR
CAYM10233-1
Anti-FABP3 Mouse Monoclonal Antibody [clone: CC68]
Vasta-aineet
Type: Primary
Antigen: FABP3
Clonality: Monoclonal
Clone: CC68
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR