Anti-SMAD3 Rabbit Polyclonal Antibody

Toimittaja: Cayman Chemical
CAYM10832-100EA 524 EUR
CAYM10832-100
Anti-SMAD3 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: SMAD3
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat, Rhesus Monkey, fish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR