Anti-SMAD5 Rabbit Polyclonal Antibody

Toimittaja: Cayman Chemical
CAYM10838-100EA 524 EUR
CAYM10838-100
Anti-SMAD5 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: SMAD5
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, New World Monkey, Mouse, Rat, Drosphila, Pig, pufferfish, Sheep, Xenopus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR