Anti-Cav-beta-4 Mouse Monoclonal Antibody [clone: S10-7]

CAYM13701-100EA 670 EUR
CAYM13701-100
Anti-Cav-beta-4 Mouse Monoclonal Antibody [clone: S10-7]
Vasta-aineet
Type: Primary
Antigen: Cav-beta-4
Clonality: Monoclonal
Clone: S10-7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR